Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009354.1 | NP_033380.1 | 3 - 4 | 1766 | 67 | |
GenBank mRNA | AF029235.1 | - | 1 - 2 | 90 | 67 | |
AF051911.1 | - | 3 - 4 | 1766 | 67 | ||
AF073311.1 | - | 3 - 4 | 1737 | 67 | ||
BC082327.1 | - | 4 - 5 | 1268 | 67 | ||
BC127068.1 | - | 4 - 5 | 1054 | 40110173 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |