Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007924.2 | NP_031950.2 | 2 - 3 | 213 | 74 | |
GenBank mRNA | AK159027.1 | - | 2 - 3 | 200 | 74 | |
BY252808.1 | - | 2 - 3 | 197 | 74 | ||
BY272058.1 | - | 2 - 3 | 214 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |