Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007924.2 | NP_031950.2 | 7 - 8 | 968 | 107 | |
GenBank mRNA | AK159027.1 | - | 7 - 8 | 955 | 107 | |
BC014816.1 | - | 7 - 8 | 938 | 4482339 | 107 | |
BC024894.1 | - | 7 - 8 | 937 | 4159557 | 107 | |
U80227.1 | - | 7 - 8 | 941 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |