Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018827.2 | NP_061297.1 | 3 - 4 | 657 | 131 | |
GenBank mRNA | AB040038.1 | - | 3 - 4 | 657 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |