Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133974.3 | NP_598735.2 | 4 - 5 | 1146 | 60 | |
GenBank mRNA | AK035674.1 | - | 4 - 5 | 1108 | 60 | |
AK087108.1 | - | 4 - 5 | 1046 | 60 | ||
BC034137.1 | - | 2 - 3 | 554 | 60 | ||
BC059937.1 | - | 4 - 5 | 1053 | 60 | ||
BC085253.1 | - | 4 - 5 | 1146 | 5347122 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |