Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027182.2 | NP_081458.1 | 2 - 3 | 446 | 90 | |
GenBank mRNA | AK010336.1 | - | 2 - 3 | 408 | 90 | |
AK142463.1 | - | 419 | 90 | |||
AK146877.1 | - | 2 - 3 | 447 | 90 | ||
AK151568.1 | - | 2 - 3 | 382 | 90 | ||
BC064315.1 | - | 2 - 3 | 369 | 90 | ||
BC126946.1 | - | 2 - 3 | 345 | 40090292 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |