Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028959.3 | NP_083235.3 | 9 - 10 | 1561 | 147 | |
GenBank mRNA | AK011607.1 | - | 4 - 5 | 485 | 147 | |
AK017134.1 | - | 9 - 10 | 1553 | 147 | ||
AK034892.1 | - | 9 - 10 | 1554 | 147 | ||
AK161033.1 | - | 5 - 6 | 886 | 147 | ||
AK173189.1 | - | 2 - 3 | 3545 | 147 | ||
BC060686.1 | - | 8 - 9 | 2549 | 147 | ||
BC080681.1 | - | 9 - 10 | 1277 | 147 | ||
BC100549.1 | - | 8 - 9 | 1411 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |