Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam81b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BB615662.1 | - | 3 - 4 | 395 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons |