Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138953.2 | NP_620403.2 | 4 - 5 | 646 | 82 | |
GenBank mRNA | AK140906.1 | - | 4 - 5 | 600 | 82 | |
AK144978.1 | - | 4 - 5 | 607 | 82 | ||
AK145868.1 | - | 4 - 5 | 595 | 82 | ||
AK154545.1 | - | 4 - 5 | 607 | 82 | ||
AK156164.1 | - | 4 - 5 | 612 | 82 | ||
BC151188.1 | - | 4 - 5 | 601 | 9088077 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |