Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042591.1 | NP_001036056.1 | 5 - 6 | 1100 | 81 | |
GenBank mRNA | AK084951.1 | - | 5 - 6 | 1111 | 81 | |
AK088288.1 | - | 5 - 6 | 1115 | 81 | ||
AK122499.1 | - | 5 - 6 | 1087 | 81 | ||
AK163364.1 | - | 5 - 6 | 645 | 81 | ||
BC023838.1 | - | 2 - 3 | 318 | 81 | ||
BC031125.1 | - | 3 - 4 | 478 | 81 | ||
BC044762.1 | - | 5 - 6 | 1098 | 81 | ||
BC054844.1 | - | 5 - 6 | 1100 | 6493441 | 81 | |
BC098474.1 | - | 5 - 6 | 1094 | 30094818 | 81 | |
DQ861994.1 | - | 5 - 6 | 845 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |