Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833411 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833410 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833407 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833408 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833409 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2174233 |
AK045944.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2174233 |
AK141135.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2174233 |
NM_011758.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
BC011182.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
BC108271.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
AK045944.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
AK141135.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
NM_011758.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
BC011182.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
BC108271.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
AK045944.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
AK141135.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P600535 |
NM_011758.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
797065 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838998 |
AK002507.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838998 |
AK007924.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838998 |
AK028605.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838998 |
BC012692.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838998 |
NM_018859.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838998 |
U68535.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
49074 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543284 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
213848 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858993 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527167 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969403 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
663393 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2671 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
146701 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P134748 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
BC011182.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
BC108271.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
AK045944.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
AK141135.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
NM_011758.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
BC011182.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
BC108271.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
AK045944.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
AK141135.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P600535 |
NM_011758.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2174233 |
AK045944.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2174233 |
AK141135.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2174233 |
NM_011758.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833439 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833418 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833435 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833404 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833401 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833403 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833402 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833442 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833407 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833405 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833415 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833414 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833413 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833412 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833440 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833395 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833398 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833406 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833437 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833410 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833396 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27833419 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P134748 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07358445 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07358422 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07358432 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P600535 |
BC011182.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
BC108271.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
AK045944.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
AK141135.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
NM_011758.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
BC011182.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
BC108271.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
AK045944.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
AK141135.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P600535 |
NM_011758.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P114472 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P106234 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P134748 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P02336326 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09352430 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08984865 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08984861 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08984862 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09200890 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833411 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833409 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833408 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833410 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27833407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833398 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833304 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833289 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833302 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833312 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833334 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833300 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833325 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833372 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833277 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833321 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27833401 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08984876 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833389 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833421 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833419 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09693197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833393 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833405 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833383 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833391 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09416758 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833397 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08984865 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833406 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833410 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09427534 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09168369 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09317584 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833401 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09520891 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833418 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833404 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833384 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833386 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09416760 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27833424 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08984862 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09427533 |
- |
- |