Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166397.1 | NP_001159869.1 | 4 - 5 | 735 | 149 | |
NM_001166398.1 | NP_001159870.1 | 4 - 5 | 678 | 149 | ||
NM_026139.4 | NP_080415.3 | 3 - 4 | 842 | 149 | ||
GenBank mRNA | AK014329.1 | - | 4 - 5 | 511 | 149 | |
AK034185.1 | - | 4 - 5 | 511 | 149 | ||
AK034621.1 | - | 4 - 5 | 509 | 149 | ||
AK043440.1 | - | 4 - 5 | 511 | 149 | ||
AK044926.1 | - | 4 - 5 | 509 | 149 | ||
AK053354.1 | - | 3 - 4 | 616 | 149 | ||
AK081299.1 | - | 4 - 5 | 454 | 149 | ||
AK172960.1 | - | 4 - 5 | 735 | 149 | ||
BC049088.1 | - | 4 - 5 | 490 | 149 | ||
BC052383.1 | - | 4 - 5 | 484 | 149 | ||
BC054839.1 | - | 1 - 2 | 74 | 6490133 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |