Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289735.1 | NP_001276664.1 | 6 - 7 | 673 | 73 | |
NM_001289736.1 | NP_001276665.1 | 6 - 7 | 615 | 73 | ||
NM_031259.2 | NP_112549.1 | 6 - 7 | 673 | 73 | ||
GenBank mRNA | AB178900.1 | - | 6 - 7 | 673 | 73 | |
AF285575.1 | - | 6 - 7 | 673 | 73 | ||
AY017476.1 | - | 6 - 7 | 615 | 73 | ||
BC137928.1 | - | 6 - 7 | 672 | 8860947 | 73 | |
BC137929.1 | - | 6 - 7 | 672 | 8860948 | 73 | |
BC145056.1 | - | 6 - 7 | 672 | 9053604 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |