Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009654.3 | NP_033784.2 | 9 - 10 | 1236 | 102 | |
GenBank mRNA | AJ011413.1 | - | 9 - 10 | 1223 | 102 | |
AJ457860.1 | - | 9 - 10 | 1190 | 102 | ||
AK010025.1 | - | 9 - 10 | 1225 | 102 | ||
AK050248.1 | - | 9 - 10 | 1225 | 102 | ||
AK050644.1 | - | 8 - 9 | 1094 | 102 | ||
AK131891.1 | - | 9 - 10 | 1228 | 102 | ||
AK160487.1 | - | 9 - 10 | 1228 | 102 | ||
BC024643.1 | - | 9 - 10 | 1210 | 4242076 | 102 | |
BC049971.1 | - | 9 - 10 | 1210 | 4161986 | 102 | |
M16111.1 | - | 6 - 7 | 896 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |