Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400014553.mm.1 |
NM_173867.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014553.mm.1 |
NM_173867.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014553.mm.1 |
TC0400001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014554.mm.1 |
NM_173867.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014554.mm.1 |
NM_173867.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014554.mm.1 |
TC0400001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014557.mm.1 |
NM_173867.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014557.mm.1 |
NM_173867.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014557.mm.1 |
TC0400001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014552.mm.1 |
NM_173867.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014552.mm.1 |
NM_173867.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014552.mm.1 |
TC0400001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014555.mm.1 |
NM_173867.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014555.mm.1 |
NM_173867.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014555.mm.1 |
TC0400001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014556.mm.1 |
NM_173867.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014556.mm.1 |
NM_173867.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014556.mm.1 |
TC0400001766.mm.1 |
MTA TC |