Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026994.1 | NP_081270.1 | 9 - 10 | 1001 | 64 | |
NM_133679.2 | NP_598440.1 | 10 - 11 | 1046 | 64 | ||
GenBank mRNA | AK145827.1 | - | 10 - 11 | 870 | 64 | |
AK170997.1 | - | 2 - 3 | 162 | 64 | ||
BC010479.1 | - | 10 - 11 | 843 | 3968727 | 64 | |
BC019387.1 | - | 9 - 10 | 810 | 4164847 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |