Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163755.1 | NP_001157227.1 | 2 - 3 | 2314 | 94 | |
NM_001163757.1 | NP_001157229.1 | 2 - 3 | 2314 | 94 | ||
NM_001163758.1 | NP_001157230.1 | 2 - 3 | 2295 | 94 | ||
NM_172446.3 | NP_766034.2 | 2 - 3 | 2367 | 94 | ||
GenBank mRNA | AB185113.1 | - | 2 - 3 | 2317 | 94 | |
AK029916.1 | - | 2 - 3 | 2298 | 94 | ||
AK049035.1 | - | 2 - 3 | 2368 | 94 | ||
AK134840.1 | - | 2 - 3 | 2306 | 94 | ||
BC138063.1 | - | 2 - 3 | 2271 | 8861082 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |