Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 4 - 5 | 777 | 89 | |
NM_019867.2 | NP_063920.2 | 2 - 3 | 414 | 89 | ||
GenBank mRNA | AJ238898.1 | - | 2 - 3 | 395 | 89 | |
AK138433.1 | - | 2 - 3 | 384 | 89 | ||
AY038025.1 | - | 2 - 3 | 389 | 89 | ||
BC039279.1 | - | 4 - 5 | 777 | 4973826 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |