Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270435.1 | NP_001257364.1 | 10 - 11 | 1706 | 78 | |
NM_001270446.1 | NP_001257375.1 | 5 - 6 | 962 | 78 | ||
NM_028506.1 | NP_082782.1 | 6 - 7 | 969 | 78 | ||
NM_029286.2 | NP_083562.1 | 10 - 11 | 1721 | 78 | ||
GenBank mRNA | AK049010.1 | - | 5 - 6 | 862 | 78 | |
AK078362.1 | - | 10 - 11 | 1648 | 78 | ||
AK163185.1 | - | 10 - 11 | 1647 | 78 | ||
AK224940.1 | - | 6 - 7 | 898 | 78 | ||
AK224961.1 | - | 5 - 6 | 961 | 78 | ||
BC114436.1 | - | 10 - 11 | 1648 | 40059415 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |