Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021604.3 | NP_067617.3 | 8 - 9 | 1676 | 80 | |
GenBank mRNA | AK147577.1 | - | 7 - 8 | 1697 | 80 | |
BC058651.1 | - | 7 - 8 | 1789 | 80 | ||
BC150703.1 | - | 6 - 7 | 1130 | 9087614 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |