Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039128.1 | - | 152 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |