Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338427 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338431 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338424 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338430 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2075479 |
AK003501.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2075479 |
AK011634.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2075479 |
NM_009084.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2075479 |
AK012655.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2075479 |
AK013246.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
903495 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160345 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957204 |
BC086796.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957204 |
BC096413.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2075479 |
AK003501.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2075479 |
AK011634.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2075479 |
NM_009084.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2075479 |
AK012655.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2075479 |
AK013246.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P441263 |
BP773503.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P441263 |
AK003501.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P441263 |
AK011634.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P441263 |
NM_009084.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P441263 |
AK012655.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338430 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338435 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338449 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338445 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338452 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338424 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338442 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338447 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338433 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338437 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338451 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20338438 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160345 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P441263 |
BP773503.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P441263 |
AK003501.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P441263 |
AK011634.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P441263 |
NM_009084.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P441263 |
AK012655.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136564 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113410 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160345 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00904376 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00904373 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00904372 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00350506 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338424 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338430 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338431 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20338427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20338448 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00693056 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338411 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338482 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338449 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338437 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338418 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338434 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338476 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00604837 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338451 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338484 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00093322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338464 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338496 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338421 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338497 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338466 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338480 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00684319 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338424 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338498 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338414 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00758182 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338492 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338427 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338422 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00906310 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338490 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338460 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338430 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338435 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00272029 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00760160 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338415 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00292563 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338478 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338445 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338447 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00684322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00760901 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00212047 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338473 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338485 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338468 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00684320 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338471 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338488 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20338494 |
- |
- |