Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470079 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470051 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470053 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470054 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470077 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470078 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK168294.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK046610.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK052819.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK089083.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK035837.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK168294.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK046610.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK052819.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK089083.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P644674 |
AK035837.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497960 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
139960 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916912 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
112153 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
418556 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914688 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
935563 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
111048 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416542 |
AK077410.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416542 |
AK147990.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416542 |
BC015081.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416542 |
BC094034.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416542 |
NM_008971.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
926556 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
5152 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
71991 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
146347 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123571 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P176545 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK168294.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK046610.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK052819.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK089083.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK035837.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK168294.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK046610.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK052819.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK089083.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P644674 |
AK035837.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470059 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470043 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470057 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470054 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470072 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470049 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470063 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470075 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470037 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470078 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470040 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470076 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470044 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470039 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470061 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470055 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470041 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470073 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470067 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470058 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470070 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470064 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470046 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28470068 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P123571 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P176545 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07635147 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07635115 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK168294.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK046610.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK052819.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK089083.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK035837.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK168294.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK046610.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK052819.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK089083.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P644674 |
AK035837.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P142981 |
AK052819.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P142981 |
AK035837.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P432354 |
AK168294.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P432354 |
BC031464.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P432354 |
AK046610.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P432354 |
AK089083.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P432354 |
NM_030057.2 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P176545 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P123571 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10162042 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10162049 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10162043 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10162045 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09908142 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09770043 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470053 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470079 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470077 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470051 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470078 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28470054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28469996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28470086 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470035 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28469979 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09770044 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470039 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470061 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10194009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09770043 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28469976 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470046 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470031 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470034 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470032 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470043 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470057 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28469978 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470058 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28469975 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28469974 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28469977 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470056 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470030 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470037 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09836365 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470064 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28470049 |
- |
- |