Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700025937.mm.1 |
TC0700002914.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025937.mm.1 |
NM_001105055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025937.mm.1 |
NM_001105055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025939.mm.1 |
TC0700002914.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025939.mm.1 |
NM_001105055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025939.mm.1 |
NM_001105055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025940.mm.1 |
TC0700002914.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025940.mm.1 |
NM_001105055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025940.mm.1 |
NM_001105055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025938.mm.1 |
TC0700002914.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025938.mm.1 |
NM_001105055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025938.mm.1 |
NM_001105055.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025941.mm.1 |
TC0700002914.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025941.mm.1 |
NM_001105055.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025941.mm.1 |
NM_001105055.1 |
Refseq |