Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012052.1 | - | 4 - 5 | 390 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |