Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025433.3 | NP_079709.2 | 4 - 5 | 445 | 132 | |
GenBank mRNA | AK005264.1 | - | 4 - 5 | 434 | 132 | |
AK007884.1 | - | 4 - 5 | 434 | 132 | ||
AK008672.1 | - | 4 - 5 | 435 | 132 | ||
AK090038.1 | - | 4 - 5 | 445 | 132 | ||
AK090082.1 | - | 4 - 5 | 445 | 132 | ||
AK152288.1 | - | 3 - 4 | 374 | 132 | ||
AK166499.1 | - | 4 - 5 | 449 | 132 | ||
BC030165.1 | - | 4 - 5 | 443 | 5027519 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |