Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289828.1 | NP_001276757.1 | 2 - 3 | 627 | 82 | |
NM_001289830.1 | NP_001276759.1 | 3 - 4 | 424 | 82 | ||
NM_001289831.1 | NP_001276760.1 | 3 - 4 | 408 | 82 | ||
NM_028016.3 | NP_082292.1 | 2 - 3 | 627 | 82 | ||
GenBank mRNA | AB126939.1 | - | 2 - 3 | 409 | 82 | |
AF507043.1 | - | 2 - 3 | 599 | 82 | ||
AK010332.1 | - | 2 - 3 | 599 | 82 | ||
AK167259.1 | - | 2 - 3 | 598 | 82 | ||
AY278951.1 | - | 2 - 3 | 476 | 82 | ||
AY455282.1 | - | 3 - 4 | 408 | 82 | ||
AY455285.1 | - | 3 - 4 | 424 | 82 | ||
BC132017.1 | - | 2 - 3 | 624 | 40130294 | 82 | |
BC137873.1 | - | 2 - 3 | 624 | 8860892 | 82 | |
BC144995.1 | - | 2 - 3 | 624 | 9053543 | 82 | |
BC144996.1 | - | 2 - 3 | 624 | 9053544 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |