Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400002124.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002124.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002149.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002149.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002149.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002149.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002152.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002152.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002152.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002152.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002153.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002153.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002153.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002153.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002142.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002142.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002142.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002142.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002145.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002145.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002145.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002145.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002128.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002128.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002128.mm.1 |
NR_029594.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002128.mm.1 |
NR_029594.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002128.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002137.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002137.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002137.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002137.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002140.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002140.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002140.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002140.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002154.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002154.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002154.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002154.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002136.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002136.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002136.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002136.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002147.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002147.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002147.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002147.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002130.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002130.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002155.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002155.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002155.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002155.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002121.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002121.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002123.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002123.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002135.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002135.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002135.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002135.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002139.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002139.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002139.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002139.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002151.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002151.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002151.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002151.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002134.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002134.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002134.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002134.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002138.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002138.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002138.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002138.mm.1 |
TC0400000349.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002146.mm.1 |
NM_001164671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002146.mm.1 |
NM_001164672.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002146.mm.1 |
NM_008298.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002146.mm.1 |
TC0400000349.mm.1 |
MTA TC |