Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166506.1 | NP_001159978.1 | 16 | 146 | ||
NM_001166507.1 | NP_001159979.1 | 17 | 146 | |||
GenBank mRNA | AK172947.1 | - | 13 | 146 | ||
BC086620.1 | - | 16 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |