Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163713.1 | NP_001157185.1 | 8 | 1155 | 141 | |
GenBank mRNA | BC060959.1 | - | 8 | 1132 | 30379757 | 141 |
BU699735.1 | - | 3 | 398 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |