Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025276.3 | NP_079552.3 | 20 - 21 | 2658 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |