Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198027.2 | NP_932144.2 | 1 - 2 | 76 | 67 | |
GenBank mRNA | AK082058.1 | - | 1 - 2 | 42 | 67 | |
BY196893.1 | - | 1 - 2 | 77 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |