Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161366.1 | NP_001154838.1 | 2 - 3 | 6313 | 91 | |
NM_001161367.1 | NP_001154839.1 | 2 - 3 | 6310 | 91 | ||
NM_198418.2 | NP_940810.2 | 2 - 3 | 6310 | 91 | ||
GenBank mRNA | AB097085.1 | - | 2 - 3 | 6310 | 91 | |
BC138353.1 | - | 3 - 4 | 5698 | 8861373 | 91 | |
BC145247.1 | - | 2 - 3 | 6279 | 91 | ||
BC145248.1 | - | 2 - 3 | 6276 | 9053800 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |