Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201255.2 | NP_957707.2 | 4 - 5 | 1022 | 79 | |
GenBank mRNA | AY497550.1 | - | 4 - 5 | 1022 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus Probe spans exons |