Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173051.2 | NP_766639.2 | 3 - 4 | 339 | 159 | |
GenBank mRNA | BC104332.2 | - | 2 - 3 | 322 | 40045807 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |