Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sval3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003952.1 | NP_001003952.1 | 2 - 3 | 189 | 143 | |
GenBank mRNA | AB098509.1 | - | 2 - 3 | 189 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Probe spans exons |