Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018236.1 | - | 936 | 71 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |