Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005525.2 | NP_001005525.1 | 4 - 5 | 519 | 68 | |
GenBank mRNA | AK005213.1 | - | 2 - 3 | 250 | 68 | |
AK012697.1 | - | 5 - 6 | 586 | 68 | ||
AK054360.1 | - | 1 - 2 | 1089 | 68 | ||
AK084745.1 | - | 5 - 6 | 566 | 68 | ||
AK166941.1 | - | 7 - 8 | 757 | 68 | ||
BC008229.1 | - | 4 - 5 | 477 | 68 | ||
BC132550.1 | - | 2 - 3 | 303 | 40130827 | 68 | |
BC132552.1 | - | 2 - 3 | 303 | 40130829 | 68 | |
BC145430.1 | - | 2 - 3 | 302 | 9053988 | 68 | |
BC145431.1 | - | 2 - 3 | 303 | 68 | ||
BC145432.1 | - | 2 - 3 | 303 | 68 | ||
CK793427.1 | - | 465 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |