Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cramp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020608.3 | NP_065633.2 | 17 - 18 | 3256 | 65 | |
GenBank mRNA | AK089393.1 | - | 17 - 18 | 3516 | 65 | |
AK129356.1 | - | 8 - 9 | 1432 | 65 | ||
AK147411.1 | - | 17 - 18 | 3257 | 65 | ||
AK147449.1 | - | 17 - 18 | 3257 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |