Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034301.1 | - | 1 | 2373 | 63 | |
AK173104.1 | - | 4 | 4090 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |