Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177964.2 | NP_001171435.2 | 3 - 4 | 287 | 130 | |
GenBank mRNA | AK003729.1 | - | 3 - 4 | 288 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |