Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177964.2 | NP_001171435.2 | 6 - 7 | 586 | 136 | |
GenBank mRNA | AK003729.1 | - | 6 - 7 | 587 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |