Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100015404.mm.1 |
NM_001177900.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015404.mm.1 |
NM_023727.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015404.mm.1 |
TC0100001860.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015405.mm.1 |
NM_001177900.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015405.mm.1 |
NM_023727.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015405.mm.1 |
TC0100001860.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015407.mm.1 |
NM_001177900.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015407.mm.1 |
NM_023727.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015407.mm.1 |
TC0100001860.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015410.mm.1 |
NM_001177900.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015410.mm.1 |
NM_023727.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015410.mm.1 |
TC0100001860.mm.1 |
MTA TC |