Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277268.1 | NP_001264197.1 | 1 - 2 | 573 | 104 | |
GenBank mRNA | AB016516.1 | - | 1 - 2 | 345 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |