Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078163.1 | - | 1 - 2 | 252 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |