Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK088060.1 | - | 1 - 2 | 219 | 128 | |
AK157730.1 | - | 1 - 2 | 125 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |