Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100002047.mm.1 |
NM_001252218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002047.mm.1 |
NM_001252218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002047.mm.1 |
NM_001252219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002047.mm.1 |
NM_001252219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002047.mm.1 |
NM_053257.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002047.mm.1 |
NM_053257.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002047.mm.1 |
TC0100000284.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002050.mm.1 |
NM_001252218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002050.mm.1 |
NM_001252218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002050.mm.1 |
NM_001252219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002050.mm.1 |
NM_001252219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002050.mm.1 |
TC0100000284.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002054.mm.1 |
NM_001252218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002054.mm.1 |
NM_001252218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002054.mm.1 |
NM_001252219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002054.mm.1 |
NM_001252219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002054.mm.1 |
NM_053257.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002054.mm.1 |
NM_053257.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002054.mm.1 |
TC0100000284.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002052.mm.1 |
NM_001252218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002052.mm.1 |
NM_001252218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002052.mm.1 |
NM_001252219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002052.mm.1 |
NM_001252219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002052.mm.1 |
NM_053257.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002052.mm.1 |
NM_053257.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002052.mm.1 |
TC0100000284.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002053.mm.1 |
NM_001252218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002053.mm.1 |
NM_001252218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002053.mm.1 |
NM_001252219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002053.mm.1 |
NM_001252219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002053.mm.1 |
NM_053257.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002053.mm.1 |
NM_053257.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002053.mm.1 |
TC0100000284.mm.1 |
MTA TC |