Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025586.3 | NP_079862.1 | 3 - 4 | 353 | 61 | |
GenBank mRNA | AK002234.1 | - | 3 - 4 | 350 | 61 | |
AK010319.1 | - | 3 - 4 | 354 | 61 | ||
AK010674.1 | - | 3 - 4 | 352 | 61 | ||
AK010916.1 | - | 3 - 4 | 354 | 61 | ||
AK010931.1 | - | 3 - 4 | 354 | 61 | ||
AK012025.1 | - | 3 - 4 | 350 | 61 | ||
AK012423.1 | - | 3 - 4 | 345 | 61 | ||
AK019386.1 | - | 2 - 3 | 341 | 61 | ||
AK151138.1 | - | 3 - 4 | 350 | 61 | ||
AK152711.1 | - | 3 - 4 | 351 | 61 | ||
AK159183.1 | - | 3 - 4 | 351 | 61 | ||
AK159626.1 | - | 3 - 4 | 351 | 61 | ||
AK167996.1 | - | 3 - 4 | 351 | 61 | ||
AK168125.1 | - | 3 - 4 | 354 | 61 | ||
AK168807.1 | - | 2 - 3 | 345 | 61 | ||
AK169662.1 | - | 3 - 4 | 351 | 61 | ||
AK186686.1 | - | 1 - 2 | 117 | 61 | ||
BC079842.1 | - | 3 - 4 | 340 | 5684456 | 61 | |
BC081441.1 | - | 2 - 3 | 337 | 6823341 | 61 | |
BC081442.1 | - | 2 - 3 | 333 | 6824136 | 61 | |
BC087917.1 | - | 3 - 4 | 380 | 30299997 | 61 | |
BC091735.1 | - | 2 - 3 | 330 | 3667351 | 61 | |
BC100335.1 | - | 2 - 3 | 320 | 30935863 | 61 | |
BC100561.1 | - | 625 | 1361804 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |