Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100013732.mm.1 |
NM_033568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013732.mm.1 |
NM_033568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013732.mm.1 |
TC1100001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100013736.mm.1 |
NM_033568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013736.mm.1 |
NM_033568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013736.mm.1 |
TC1100001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100013740.mm.1 |
NM_033568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013740.mm.1 |
NM_033568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013740.mm.1 |
TC1100001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100013737.mm.1 |
NM_033568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013737.mm.1 |
NM_033568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013737.mm.1 |
TC1100001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100013738.mm.1 |
NM_033568.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013738.mm.1 |
NM_033568.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100013738.mm.1 |
TC1100001482.mm.1 |
MTA TC |