Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445383 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445382 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445385 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445381 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445384 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445379 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445380 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2080603 |
AK219581.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2080603 |
AK217841.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2080603 |
AK192871.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2080603 |
AK202523.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2080603 |
AK205012.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2069721 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826581 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463729 |
AB257853.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463729 |
AK033439.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463729 |
NM_001081017.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
18660 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2080603 |
AK219581.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2080603 |
AK217841.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2080603 |
AK192871.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2080603 |
AK202523.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2080603 |
AK205012.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2069721 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P170371 |
AK219581.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P170371 |
AK192871.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P170371 |
M19141.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P170371 |
U27129.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P170371 |
BC106193.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445371 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445360 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445380 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445383 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445362 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445403 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445373 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445384 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445387 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445402 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445351 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445369 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445388 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445365 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445391 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445361 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445352 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445354 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445358 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445357 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445386 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445381 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445355 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445367 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445353 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445374 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445394 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445392 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26445379 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06759973 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P170371 |
AK219581.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P170371 |
AK192871.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P170371 |
M19141.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P170371 |
U27129.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P170371 |
BC106193.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
U27129.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
BC106193.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
X05837.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
BC089457.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P12877 |
BC094900.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115095 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133783 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06759973 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07655114 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07655120 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445384 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445381 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445380 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445383 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445385 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445382 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26445379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26445429 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23457440 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23457437 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23457438 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554331 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445412 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08122447 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554345 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08165500 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445371 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445419 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08038402 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08148696 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05699379 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445354 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07915960 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445357 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445361 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554352 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554354 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445391 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445367 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445411 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445344 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554347 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445359 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445369 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445373 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445364 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445378 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445386 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445383 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445348 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445403 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445406 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08122438 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554350 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554334 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445432 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554332 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554342 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554339 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445358 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445346 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554344 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445388 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445440 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05083242 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07974328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445392 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07974327 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07941072 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445433 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07750793 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554330 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08165501 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445402 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445396 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445438 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07675104 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554336 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445409 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445430 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445420 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05257184 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08122443 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24554353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445441 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445355 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445435 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445405 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445417 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445443 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445362 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26445350 |
- |
- |